Masterflex® Ismatec® Pump Tubing, Tygon® LMT-55, 9.5 mm ID x 15.9 mm OD; 50 Ft
Supplier: VWR International
VWR Item#MFLX06507-39
Your Organization's Price:
Retrieving
Exceptional durability with long life.
  • Crystal clear and flexible while also being long-lasting and crack-resistant
  • Dimensions: 9.5 mm ID x 15.9 mm OD
Applications: General laboratory, food and beverage, biopharmaceutical, analytical instruments<br /><br />Sterilize by autoclave or ethylene oxide gas.