Masterflex® I/P® Precision Pump Tubing, Puri-Flex®, I/P 82; 25 ft
Supplier: VWR International
VWR Item#MFLX96419-82
Your Organization's Price:
Retrieving
High-performance and long-life tubing for high-purity applications.
  • Genuine Masterflex® tubing undergoes robust testing to provide optimal performance/pump life hours with peristaltic pump equipment
  • Precision extruded to tight tolerances for high repeatability, accuracy, and reproducibility
  • Single-use custom lengths and configurations available to minimize downtime
  • Low spallation when compared to silicone or some other TPE tubings.
  • I/P® 82 tubing has a 1/2” (12.7 mm) inner diameter
  • Accepts 1/2” hose barb for tubing connection
  • Certificate of compliance included with each individual unit upon shipment
Masterflex® Puri-Flex™ Precision pump tubing contains no DEHP phthalates or halogen plasticizers to protect fluid purity. The soft and flexible material is heat sealable and weldable and transparent for easy flow monitoring.
This tubing features ultralow extractables or leachables and has low spallation to the fluid. Ideal for cell and tissue culture work, biotech, and pharmaceutical applications, this tubing complies with USP Class VI, REACH, and RoHS standards.