Masterflex® Transfer Tubing, PVDF, 3/8" ID x 1/2" OD; 25 Ft
Supplier: Saint Gobain Life Sciences
VWR Item#MFLX95100-10
Your Organization's Price:
Retrieving
Fluoropolymer tubing at less cost.
  • Ideal for general-purpose, single-use, and extended fluid path applications
  • Single-use custom lengths and configurations available to minimize downtime
  • Tubing size: 3/4" ID x 7/8" OD
This tubing is ideal for high-purity applications and corrosive chemical transfer—an inexpensive alternative to fluoropolymer tubing. The polyvinylidene fluoride (PVDF) material is kink resistant, has excellent chemical resistance, and is odorless and nontoxic. The resin is FDA-compliant (21 CFR 177.2600); meets USP 23 Class VI and USDA requirements. It has a temperature range of -40 to 260 °F (-40 to 127 °C). Can be <br />sterilized with autoclave and gamma irradiation.