LMI Digital Remote-Control Solenoid-Diaphragm Metering Pump, 2.5 GPH, 115 VAC
Supplier: EQUIP SOLUTIONS
VWR Item#MFLX74516-20
Your Organization's Price:
Retrieving
Manually adjustable stroke length and stroke frequency provide precise control of flow rates.
  • Clear face cover protects pump controls on low flow models (1 GPH and lower)
  • Chemical-resistant housing provides protection in harsh environments
Microprocessor control and an LCD display provides precise accuracy. Programmable controller features three external control modes — pulse multiply, pulse divide, or 4 to 20 mA response. Pumps allow you to multiply or divide incoming pulse signal by 1 to 999.<br /><br /><B>Service kits include</B> a replacement diaphragm, seal, four pump head screws, and check valves.
<b>Delivery information: </b>Includes four-function valve, 16-ft polyethylene tubing, foot valve/strainer, and 8-ft power cord with molded plug.