Micropump® L21262 A-Mount Cavity Style Pump Head; SS, PEEK, PTFE; 0.32 mL/rev
Supplier: MICROPUMP INC
Smooth pulseless flow provides precise, leak-free delivery of fluids.
- Cavity-style pump heads are designed to handle higher inlet pressures.
- Available pump head service kits include: gears, suction shoe, shaft seals, and springs.
- Call our Application Specialists for assistance.
Magnetically driven, precision-geared pump heads provide smooth, accurate, pulseless fluid delivery. The magnetic drive and static seals keep fluids securely inside and potential contaminants out. Chemical resistant materials handle a wide range of materials (due to wetted parts). Pump heads are compact and easy to service. Modular design offers flexibility—configure to your specific needs.
Available pump head service kits include: gears, suction shoe, shaft seals, and springs.<br />Call our Application Specialists for assistance.