GF Signet 3-9900 Batch Module for Flow Transmitter 56560-12
Supplier: BURT PROCESS EQUIPMENT
Optimize performance of your GF+ Signet Transmitter/Controller.
- Choose from a wide range of replacement parts and accessories
Use modules, sensors, and RC filters to expand the controller's capabilities. Adapt your controller to your existing system with a variety of mounting options.
Should be used in conjunction with a 56560-12 or 56560-14 9900 Series Transmitter, and 56560-15 relay module in order to create a Batch Controller System.