Cadence Luer Fitting Adapter, Nickel-Plated Brass, Male Luer Lock to 10-32 UNF
Supplier: CADENCE INC.
VWR Item#MFLX41507-70
Your Organization's Price:
Retrieving
Excellent durability.
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimize downtime
  • Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
  • Manufactured in an ISO 14644-1 Level 8 (formerly Class 100,000) clean room
  • Full material traceability on all items
  • Connection Type: Luer Adapter, Female Luer Lock x Plug
Constructed of durable plated brass, these Luer Adapter Fittings are ideal for a wide range of applications.