Cadence Luer Stopcock Fitting, Nickel-Plated Brass, Three-Way, Male Luer Lock x Male Luer Lock x Side-Arm Female Luer
Supplier: CADENCE INC.
VWR Item#MFLX41507-13
Your Organization's Price:
Retrieving
Exceptional durability.
  • Easy to disassemble for cleaning
  • Stopcock handles turn 180°
  • Autoclavable
These Cadence Science luer stopcocks are constructed of nickel-plated brass for enhanced durability. These stopcocks offer worry-free cleaning with easy disassembly to save valuable time.