Fitting, PTFE, Straight, Hose Barb to Threaded Adapter, 1/8" NPT(F)×1/4" ID
Supplier: MICROMOLD PRODUCTS, INC.
VWR Item#MFLX34030-89
Your Organization's Price:
Retrieving
No tools needed for assembly.
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimize downtime
  • Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
Barbed fittings are designed for quick and easy assembly without tools. Combine these simple and economical fittings with our flexible tubing for use in your low-pressure applications (generally less than 30 psi).