Female Threaded Tee Coupling, 1/4" NPT(F), PTFE
Supplier: MICROMOLD PRODUCTS, INC.
Maintain a permanent connection.
- Ideal for general-purpose and single-use applications, to connect tubing with rest of system
- Determine which connector type is needed for your application by contacting free technical application assistance*
- Global market leader in range of materials for most extensive chemical compatibility
- Single-use custom configuration solutions to minimize downtime
- Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
- Connection Type: All three ports ³/₄" NPT(F)
Threaded fittings are designed for more permanent installations. Choose the material and size that meets your application needs.