McMillan Flow Modular System Flow Sensor, Ryton® PPS, for Liquids, 100 to 1000 mL/min
Supplier: MC MILLAN CO
VWR Item#MFLX32703-54
Your Organization's Price:
Retrieving
Ryton® PPS materials for use in aggressive and non-aggressive gas or liquid systems.
  • A range of available configurations to suit most applications
  • Voltage output to monitor and record flow rates and totals
  • Interface with data acquisition system
These sensors are ideal for low-flow applications involving mildly acidic or slightly corrosive gases and liquids. Economically designed, sensors provide a single 0 to 5 VDC output signal. This single signal is ideal for simple, low-cost flow rate measurement or for integration of the sensor into an existing central control system.<br /><br />Ryton PPS sensors are ideal for liquids or air. Sensors for liquids can be used with a wide variety of transparent, low-viscosity liquids under 10 cSt. Sensors measure a wide flow range from as low as 20 ml/min to as high as 500 L/min.<br /><br />The wetted materials are epoxy, glass-filled polyphenylene sulfide (Ryton PPS), glass, stainless steel, sapphire, and Viton.<br /><br /><B>Note:</B> A power supply is required for these flow sensors—order separately. If using an existing power supply, order cable assembly 32704-52.