Hedland H701A-030 Oil and Petroleum Flowmeter, 3-30 GPM, Al, 3/4"
Supplier: BADGER METER INC
VWR Item#MFLX32508-37
Your Organization's Price:
Retrieving
Install in any position—360° rotatable guard/scale.
  • Easier to read linear scale
<B> FEATURES</B><br />Direct reading<br />Install in any posiiton<br />360 degrees rotatable guard / scale<br />Easier to read linear scale<br />No flow straighteners or special piping required<br />Relatively insensitive to shock and vibration<br />Good viscosity stability<br />Temperature up to 240 °F<br />Accuracy ± 2% full scale<br />Repeatability ± 1%<br />Special scales available