Masterflex® Fitting, Polypropylene, 90° Elbow, Sanitary Clamp Union, 3/4" Clamp x 3/4" Clamp
Supplier: VWR International
VWR Item#MFLX31806-41
Your Organization's Price:
Retrieving
Maintain sanitary conditions while securely connecting sanitary process lines.
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimize downtime
  • Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
  • Meet USP Class VI for pharmaceutical use and FDA, USDA, and 3A standards for food contact
  • All fittings are compatible with Tri-Clamp® fittings
  • Excellent chemical resistance; low leachable characteristics
  • Connection Type: 3/4" Clamp x 3/4" Clamp
Tri-Clamp® 90-degree elbow adapters allow for secure connection of sanitary lines when a 90-degree angle is required.<br /><br />Though both materials may be autoclaved, PVDF tolerates high temperatures better than PP does and will perform more consistently after repeated autoclaving.<br /><br />Clamps and gaskets required for a complete system are sold separately.