Masterflex® Fitting, Polypropylene, Straight, Hose Barb Reducer, 1" ID x 1/2" ID; 10/PK
Supplier: VWR International
Best for low-pressure applications.
- Ideal for general-purpose and single-use applications, to connect tubing with rest of system
- Determine which connector type is needed for your application by contacting free technical application assistance*
- Global market leader in range of materials for most extensive chemical compatibility
- Single-use custom configuration solutions to minimize downtime
- Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
- Temperture range 9 to 180 °F (-13 to 66 °C)
- Connection Type: 1" ID x 1/2" ID