Masterflex® Fitting, Polypropylene, Straight, Hose Barb Reducer, 3/4" ID x 1/2" ID; 10/PK
Supplier: VWR International
VWR Item#MFLX30616-71
Your Organization's Price:
Retrieving
Best for low-pressure applications.
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimize downtime
  • Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
  • Temperture range 9 to 180 °F (-13 to 66 °C)
  • Connection Type: 3/4" ID x 1/2" ID