PendoTech TT1 Single-Use Temperature Sensor Transmitter with 4-20 mA Output
Supplier: PENDOTECH
VWR Item#MFLX19406-45
Your Organization's Price:
Retrieving
Optimize performance of your PendoTECH® Pressure Sensor.
  • Choose from a wide range of replacement parts and accessories
  • Produces a 4-20 milliamp signal that is linear with temperature
  • Connections to the transmitter are quick and convenient
  • Convenient mounting screws
Replacement parts and accessories to keep your PendoTECH systems running smoothly and accurately.
The temperature sensor transmitter easily connects to the PendoTECH temperature sensor. The narrow 4-20mA output range from 0 to 70ºC ensures accurate and drift free linearization of the sensor output.
***Must order reusable temperature sensor cables (19406-14 for luer sensor and 19406-15 for hose barb sensors) separately.***
<b>Delivery information: </b>Power cord and plug.