Flux Pumps Air-Powered Drum Pump Motor, 1/2 HP, 9000 rpm; 17 cfm@85 psi
Supplier: FLUX PUMPS CORP
VWR Item#MFLX07070-60
Your Organization's Price:
Retrieving
Configurable to your requirements.
  • High-viscosity tube set handles fluids with viscosities to 50,000 cp!
Air-operated motors are ideal for hazardous duty applications or where electricity is not available. Models 07070-60 and -70 are fitted with a muffler and include a grounding post. Model 07070-70 also has a unique trigger valve handle—press switch for intermittent pumping; lock switch for continuous use.
<B>Note:</B> This motor can only be used with tube sets 70603-30, -32, -34, -36, -40, -42, -44 and -46.