Fitting, PFA, Elbow, Threaded Union, 1/2" NPT(F)
Supplier: ANDON SPECIALTIES INC.
Chemically-inert PFA handles most aggressive chemicals.
- Quick and easy assembly without tools
Threaded fittings offer a more permanent installation. Choose the size to meet your application needs.