Lever Drum Pump, PP/SS/Viton, 8/10/12 oz/Stroke; 2" IPS Bung
Supplier: Action Pump
VWR Item#MFLX06511-50
Your Organization's Price:
Retrieving
Economical pumps for use in cabinets or tight work spaces.
  • Fit 15, 30, and 55-gallon drums with 2" IPS bung openings
  • Designed for use in industrial, agricultural, labs, and automotive industries
Choose from center-level pump and lever-action pump model options.<br /><br />Center-Lever Pump is self-priming and features an anti-drip spout to eliminate wasted fluid. This pump is ideal for oils, detergents, and other compatible industrial fluids.<br /><br />Lever-Action Pumps feature a curved discharge spout for convenient hose extension. Choose from three models depending on your chemical needs. Model 06511-50 is ideal for water, alcohol, gasoline, diesel, kerosene, and lubricants. Model 06511-60 handles the same fluids, plus water-based chemicals, acids, and alkalis. Model 06511-80 handles the same fluids, plus strong acids and strong alkalis.
Lever-Action Pumps feature a curved discharge spout or convenient hose extension. Choose from three models depending on your chemical needs. Model 06511-50 is ideal for water, alcohol, gasoline, diesel, kerosene, and lubricants. Model 06511-60 handles the same fluids, plus water-based chemicals, acid, and alkaline. Model 06511-80 handles the same fluids, plus strong acids and strong alkalis.