Masterflex® Ismatec® Pump Tubing, Tygon® LMT-55, 3.2 mm ID x 6.4 mm OD; 50 Ft
Supplier: VWR International
Exceptional durability with long life.
- Crystal clear and flexible while also being long-lasting and crack-resistant
- Dimensions: 3.2 mm ID x 6.4 mm OD
Applications: General laboratory, food and beverage, biopharmaceutical, analytical instruments<br /><br />Sterilize by autoclave or ethylene oxide gas.