Double open end semiconductor filter cartridge; 0.20 µm
Supplier: HARRINGTON INDUSTRIAL PLASTICS LLC
VWR Item#MFLX06479-60
Your Organization's Price:
Retrieving
Ideal for ultrapure filtration applications.
  • U.S. CFR Title 21 compliant
These cartridges are autoclavable to 250 °F <121 °C) and in-line sterilizable to 257 °F (125 °C) for up to 30-minute cycles. They are tolerant to an accumulated exposure of 10 hours. The filters may also be sanitized with compatible chemical agents. All cartridges are suitable to 180 °F (82 °C) at 10 psid.<br /><br />The cartridges comply with U.S. CFR Title 21 guidelines for repeated food contact, comply with USP Class VI, 121 °C Plastics guidelines, and pass the MEM Elution Cytotoxicity Test. Aqueous extracts contain less than 0.25 EU/ml.
<B>Semiconductor Conductor</B> Single-layer PES membrane over PP core. Each is flushed with high-purity water to leach less than 5 ppb TOC and less that 5 ppb trace metals. Ideal for any high-purity chemical or DI water application.