Viton Transfer Pump Tubing, FDA-Compliant Viton®, White, 1/8" ID×3/16" OD; 25 ft
Supplier: VWR International
FDA compliant for food contact.
<B>Applications:</B> FDA compliant food or laboratory applications, acid and solvent transfer, high temperature work. <br /><B>Characteristics:</B> Good chemical resistance, opaque white. <br /><B>Certification:</B> FDA compliant. <br /><B>Temperature range:</B> -25 to 400 °F(-32 to 204 °C) <br /><B>Sterilize:</B> Circulating hot air oven at 480 °F (249 °C)