Masterflex® Transfer Tubing, PFA, 3/16" ID x 1/4" OD; 25 ft
Supplier: VWR International
VWR Item#MFLX06371-03
Your Organization's Price:
Retrieving
Stands up to highly corrosive materials.
  • Ideal for general-purpose, single-use, and extended fluid path applications
  • Single-use custom lengths and configurations available to minimize downtime
  • Tubing size: 3/16" ID x 1/4" OD
Applications: Demanding lab, chemical, electrical, mechanical, pharmaceutical, and automotive applications. For the transfer of high-temperature or highly corrosive materials. Also good for high-purity work.<br />Characteristics: Chemically inert, nontoxic, and virtually nonporous. Excellent crack- and stress-resistance. Low coefficient of friction. Translucent.<br />Resin certification: Resin is FDA-compliant (21 CFR 177.1550); meets USP Class VI requirements. Tubing is manufactured according to GMP.<br />Temperature range: -320 to 500 °F (-195 to 260 °C)<br />Sterilize: Autoclave, ethylene oxide, or use dry heat.