Masterflex® Transfer Tubing, High-Purity PFA-450, 3/8" ID x 1/2" OD; 25 ft
Supplier: VWR International
VWR Item#MFLX06368-56
Your Organization's Price:
Retrieving
Won't crack under stress.
  • Ideal for general-purpose, single-use, and extended fluid path applications
  • Single-use custom lengths and configurations available to minimize downtime
  • Tubing size:3/8" ID x 1/2" OD
<B>Applications:</B> Pharmaceutical, biotechnology, environmental, semiconductor processing, food and beverage, autoanalyzer, and nuclear applications.<br /><B>Characteristics:</B> Superior surface smoothness and clarity, low extractables with deionized water and acids, high resistance to stress and cracking. Translucent.<br /><B>Resin certification:</B> Resin is FDA compliant. <br />Tubing is manufactured according to GMP.<br /><B>Temperature range:</B> -320 to 500 °F (-195 to 260 °C)<br /><B>Sterilize:</B> With autoclave, ethylene oxide, gamma irradiation, and using dry heat.