Burkle Barbed Tee Connector, PVDF, 5-7 mm Tubing; 10/Pk
Supplier: BURKLE INC
VWR Item#MFLX06044-09
Your Organization's Price:
Retrieving
Safe and flexible connection for your most demanding applications.
  • Withstands temperatures up to 320 °F (160 °C)
  • Highly resistant to chemicals and UV
  • Autoclavable
The cylindrical barbed tee connectors are available in a variety of sizes to extend, connect, branch, join, and repair most plastic tubings. The polyvinylidene fluoride (PVDF) connectors are designed for quick and easy assembly without any tools.