Masterflex® 155394-03 Polypropylene felt filter bag, 18-1/2"L, 100 µm
Supplier: Avantor
Conveniently collect all filtered particles within the bag.
  • Choose from a wide range of bag materials suitable for your application
Choose from standard felt bags, high-efficiency bags, or nylon mesh bags. Each bag measures 4" diameter. The 10"L bags have a surface area of 0.75 ft<sup>2</sup>; 18-½"L bags have a surface area of 1.75 ft<sup>2</sup>.