Masterflex® Ball valve, 3-way, 1/8" NPT(F) - PVC w/Viton® seals
Supplier: SPECIALTY MFG CO
Nonmetallic wetted parts ideal for water and mild aqueous solutions.
- Maximum pressure: 125 psi (8.6 bar)
- Maximum temperature: 140 °F (60 °C)
Ball valves are designed primarily for on/off service.
Warning: This product is not approved or intended for, and should not be used for medical, clinical, surgical or other patient oriented applications.