Zinc ≥98%, granules, GR ACS 20 Mesh, Supelco®

EM-ZX0010-7EA 2494 USD
EM-ZX0010-7
Zinc ≥98%, granules, GR ACS 20 Mesh, Supelco®
Zinc

Meets ACS Specifications, Meets Reagent Specifications for testing USP/NF monographs

Formula: Zn
MW: 65.39 g/mol
Boiling Pt: 908
Melting Pt: 420
Density: 7.14 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011291
CAS Number: 7440-66-6
EINECS: 231-175-3
Merck Index: 12,10255

Order Now

Specification Test Results

Assay (complexometric) 99.8% min
Color Silver metallic
Form Granular
Iron (Fe) 0.01% max
Lead (Pb) 0.01% max
Mesh size (18-40) 70% min
Suitability for determination of arsenic (As) To pass test (limit 0.1 ppm)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR