VWR® Cryovials
Supplier: VWR International
VWRI210-000001 VWRI210-000003 VWRI210-000011 VWRI210-000002 VWRI210-000004 VWRI210-000010 VWRI210-000014 VWRI210-000006 VWRI210-000009 VWRI210-000005 VWRI210-000012 VWRI210-000013 VWRI210-000019 VWRI210-000007 VWRI210-000008 VWRI210-000017 VWRI210-000020 VWRI210-000016 VWRI210-000018 VWRI210-000021 VWRI210-000026 VWRI210-000027 VWRI210-000028 VWRI210-000029 VWRI210-000030 VWRI210-000015
VWRI210-000001CS 423.52 USD
VWRI210-000001 VWRI210-000003 VWRI210-000011 VWRI210-000002 VWRI210-000004 VWRI210-000010 VWRI210-000014 VWRI210-000006 VWRI210-000009 VWRI210-000005 VWRI210-000012 VWRI210-000013 VWRI210-000019 VWRI210-000007 VWRI210-000008 VWRI210-000017 VWRI210-000020 VWRI210-000016 VWRI210-000018 VWRI210-000021 VWRI210-000026 VWRI210-000027 VWRI210-000028 VWRI210-000029 VWRI210-000030 VWRI210-000015
VWR® Cryovials
Vials
Designed for freezing samples in all cryopreservation applications, these vials feature an internal thread with washer, external thread. The caps and vials are made from the same batch of materials to ensure equal dilation coefficient, resulting in optimum sealing characteristics.
  • Manufactured from polypropylene to withstand temperatures down to –196 °C
  • Manufactured in cleanroom environment (Class 10,000)
  • Free of RNase, DNase and pyrogens
  • White graduations, write on area on vial for easy marking
  • Internal vials have a washer for a secure seal
  • 2D coded vial
More Product Information