VWR® Modular Workstations, W×D×H 48×30×30"
Supplier: VWR International
VF-B483030E VF-B483030EB VF-B483030F VF-B483030L VF-B483030P VF-B483030PB
VF-B483030EEA 3026.11 USD
VF-B483030E VF-B483030EB VF-B483030F VF-B483030L VF-B483030P VF-B483030PB
VWR® Modular Workstations, W×D×H 48×30×30"
Furniture Tables and Workbenches
VWR® modular work benches are designed for a wide range of applications in laboratory and industrial environments including biotech, pharmaceutical, analytical, material handling, distribution, and assembly.
  • Can be custom configured
  • Shipped fully assembled
  • Includes a variety of attachments
The VWR® Basic Bench includes 4 standard leveler bench feet, gussets and light neutral powder coat of 16 gauge 1.5x1.5” steel frame. Shipped fully assembled. Also available in 60" and 72" widths.
Standard work benches handle 1000lbs. of evenly distributed weightMore Product Information