Horizontal Laminar Flow Clean Benches, Envirco®
Supplier: Envirco
10552 10553 10555
VF-10552EA 23162.91 USD
VF-10552 VF-10553 VF-10555
Horizontal Laminar Flow Clean Benches, Envirco®
Fume Hoods and Enclosures Laminar Flow Cabinets
The two-piece design eliminates nearly all vibration transmission from filter/blower console to work surface
The workstation frame is made of welded steel with a corrosion-resistant polyurethane enamel finish. The work surface has a depth of 61cm (24") and is 76.2Hcm (30") from floor. Benches feature removable acrylic side panels and a convenient recessed knee space for operator comfort. A duplex receptacle is located on the front leg of the bench for easy access. The nonglare, plastic laminated work surface is easy to clean. A recessed fluorescent light fixture provides 150 foot-candlesMore Product Information