AlphaSat® Vectra® Alpha® 10 Wiper, Texwipe®
Supplier: Texwipe
TX8410
TWTX8410CS 1392.26 USD
TWTX8410
AlphaSat® Vectra® Alpha® 10 Wiper, Texwipe®
Wipes Cleanroom Wipes Presaturated Cleanroom Wipes
Ultraclean wipers presaturated with 100% semiconductor-grade isopropyl alcohol.
Wipers comply with FDA and RoHS requirements. Bag-to-bag traceability.
Wipers are Vectra-processed, 100% continuous-filament double-knit polyester with thermally sealed borders. Superior strength guards against abrasion under rigorous use. Excellent for cleaning surfaces sensitive to water contamination, such as vacuum process chambers for etching and chemical vapor deposition (CVD) equipment. Suitable for maintenance of manufacturing equipment for semiconductor, disk media, and flat panels. Ideal forMore Product Information