VersaWipe® Wipers, Texwipe®
Supplier: Texwipe
TX622 TX629
TWTX622CS 309.81 USD
TWTX622 TW-TX629
VersaWipe® Wipers, Texwipe®
Wipes Cleanroom Wipes Dry Cleanroom Wipes
Wipers are suitable for cleaning precision components and lab apparatus — ideal for general wiping and spill control in controlled environments.
  • High wet and dry strength due to polyester component
  • Low particle counts, low extractable levels, and good solvent compatibility
  • No chemical binders
Made of a 55% cellulose, 45% polyester blend, these wipers combine the high sorbency of natural fiber with the cleanliness and strength of a synthetic wiper. Non-woven, hydroentangled construction offers excellent bidirectional strength.
Ordering information: More Product Information