Dimethyl sulfoxide-[D₆] 99.8 atom % D for NMR spectroscopy

Supplier: Thermo Scientific Chemicals

Synonyms: Dimethyl sulphoxide-[²H₆], Hexadeuterodimethyl sulfoxide, Hexadeuterodimethyl sulphoxide, DMSO-D6, DMSO-d6, Dimethyl sulfoxide-[²H₆], Dimethyl sulphoxide-d6, Dimethyl sulfoxide-d6

464010075
TS464010075EA 41 USD
TS464010075
Dimethyl sulfoxide-[D₆] 99.8 atom % D for NMR spectroscopy
Dimethyl sulphoxide-[D6]
Formula: D₃CS(O)CD₃
MDL Number: MFCD00002090
CAS Number: 2206-27-1
UN: 1993

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR