L-Theanine 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: (S)-2-Amino-5-(ethylamino)-5-oxopentanoic acid, L-gamma-Glutamylethylamide, N-gamma-Ethyl-L-glutamine

45934-0010 45934-0050
TS45934-0010EA 75 USD
TS45934-0010 TS45934-0050
L-Theanine 98%
L-Theanine
Formula: C₇H₁₄N₂O₃
Melting Pt: 206…219 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00059653
CAS Number: 3081-61-6

Order Now

Specification Test Results

Appearance white to off-white Crystalline powder
Infrared spectrum Conforms
Specific optical rotation +7.5° to +8.5° (20°C, 589 nm) (1 % in water)
Titration with HClO4 ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR