SurePac™ Bio 550 SEC MDi™ HPLC Columns
Supplier: Thermo Scientific
43903-032131 43903-034631 43903-037831 43903-152131 43903-154631 43903-157831
TS43903-032131EA 587.1 USD
TS43903-032131 TS43903-034631 TS43903-037831 TS43903-152131 TS43903-154631 TS43903-157831
SurePac™ Bio 550 SEC MDi™ HPLC Columns
Chromatography Columns
Experience the future of Size-Exclusion Chromatography (SEC) with SurePac™ Bio 550 SEC MDi™ analytical columns and guards exclusively designed to deliver robust and reproducible performance at high resolution and throughput to accelerate your therapeutics analysis process.
  • Ideal for AAV gene therapy characterization and quality control assessment due to selectivity of the dedicated smaller particle (3 μm) combined with wide-pore (550 Å) size
  • Superior column-to-column and lot-to-lot reproducibility eliminates column variability as a concern in stability and
More Product Information