Potassium hydroxide 1N ethanol, AcroSeal®

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Potash caustic, Potassium hydrate, Caustic potash

429091000
TS429091000EA 34 USD
TS429091000
Potassium hydroxide 1N ethanol, AcroSeal®
Potassium hydroxide
Formula: KOH
MW: 56.11 g/mol
Boiling Pt: 1320
Melting Pt: 360
Density: 0.86 g/cm³
Flash Pt: 12 °C
Storage Temperature: Ambient
MDL Number: MFCD00003553
CAS Number: 1310-58-3
UN: 2924
ADR: 3 II
Merck Index: 12,07806

Order Now

Specification Test Results

Appearance Clear colorless to yellow Solution (May darken on storage)
Strength 0.950 to 1.050 N
Melting point 179 to 183 °C
Titration with HClO4 ≥98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR