Boranepyridine complex ∼9.4 M 95%, AcroSeal™

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Pyridine Borane, Borane, pyridine complex

42713-1000
TS42713-1000EA 188 USD
TS42713-1000
Boranepyridine complex ∼9.4 M 95%, AcroSeal™
Boranepyridine complex
Formula: C₅H₈BN
Melting Pt: 10…11 °C
Density: 0.92 g/cm³
Flash Pt: 73 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00012435
CAS Number: 110-51-0
UN: 2810
ADR: 6.1,II

Order Now

Specification Test Results

Appearance Clear to slightly hazy colorless to amber Liquid (May darken on storage)
Infrared spectrum Conforms
Titration ≥94 % (Active hydrogen)
Titration with NaOH ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR