4,4'-Bi-o-toluidine dihydrochloride 99%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: o-Toluidine dihydrochloride, o-Tolidine dihydrochloride

42113-0025 42113-0250 42113-1000 42113-5000
TS42113-0025EA 1280 USD
TS42113-0025 TS42113-0250 TS42113-1000 TS42113-5000
4,4'-Bi-o-toluidine dihydrochloride 99%
4,4'-Bi-o-toluidine dihydrochloride
Formula: C₁₄H₁₈Cl₂N₂
Melting Pt: 130 °C
Storage Temperature: Ambient
MDL Number: MFCD00012960
CAS Number: 612-82-8
EINECS: 210-322-5
UN: 3077
ADR: 9,III

Order Now

Specification Test Results

Appearance Light beige to light gray Crystalline powder
Infrared spectrum Conforms
Titration Argentometric ≥98.5 % (on dry substance)
Water ≤5 % (K.F.)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR