Tin ACS 30 mesh

Supplier: Thermo Scientific Chemicals

420995000 420991000
TS420995000EA 267 USD
TS420995000 TS420991000
Tin ACS 30 mesh
Tin
Formula: Sn
Boiling Pt: 2270 °C (760 mmHg)
Melting Pt: 231.9 °C
Storage Temperature: Ambient
MDL Number: MFCD00133862
CAS Number: 7440-31-5
EINECS: 231-141-8

Order Now

Specification Test Results

Appearance Grey Metallic granular powder
Titration Complexometric ≥99.5 %
Antimony (Sb) ≤200 ppm
Arsenic (As) ≤1 ppm
Copper (Cu) ≤50 ppm
Iron (Fe) ≤100 ppm
Lead (Pb) ≤50 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR