Acetonitrile-[D₃] 99.9 atom % D for NMR spectroscopy

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Cyanomethane-D3, Acetonitrile-d3, Ethannitrile-D3, Trideuteroacetonitrile, Methyl-d3 cyanide, Methyl cyanide-D3, Acetonitrile-[²H₃]

351390100
TS351390100EA 86 USD
TS351390100
Acetonitrile-[D₃] 99.9 atom % D for NMR spectroscopy
Acetonitrile-[D3]
Formula: D₃CCN
MW: 44.03 g/mol
Boiling Pt: 80.7
Melting Pt: -42
Density: 0.844 g/cm³ (20 °C)
Flash Pt: 2 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00001881
CAS Number: 2206-26-0
EINECS: 218-616-5
UN: 1648
ADR: 3 II

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Water ≤0.03 % (Coulometric)
D-Enrichment ≥99.85 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR