Diphosphorus pentoxide 98+% ACS

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Phosphorus(V) oxide, di-Phosphorus pentoxide, Phosphorus pentoxide, Phosphoric anhydride

AC31582-0010 AC31582-2500
TS31582-0010EA 226 USD
TS31582-0010 TS31582-2500
Diphosphorus pentoxide 98+% ACS
di-Phosphorus pentoxide
Formula: P₂O₅
Melting Pt: 340…360 °C (subl.)
Storage Temperature: Ambient
MDL Number: MFCD00011440
CAS Number: 1314-56-3
EINECS: 215-236-1
UN: 1807
ADR: 8,II
Merck Index: 13,07441

Order Now

Specification Test Results

Appearance White Powder
Titration with NaOH ≥98.0 %
Ammonium (NH4) ≤100 ppm
Heavy metals (as Pb) ≤100 ppm
Phosphorus trioxide (P2O3) Passes test (limit about 0.02 %)
Insoluble matter ≤0.02 %

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR