Ethylene glycol 99.5% for analysis

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Monoethylene glycol, Ethane-1,2-diol, 2-Hydroxyethanol, Ethylene alcohol, Ethanediol, Glycol alcohol, 1,2-Dihydroxy ethane, 1,2-Ethanediol, 1,2-Dihydroxyethane, Glycol, Dihydroxyethane

29553-0010 29553-0025 29553-0250
TS29553-0010EA 119 USD
TS29553-0010 TS29553-0025 TS29553-0250
Ethylene glycol 99.5% for analysis
Ethylene glycol
Formula: HOCH₂CH₂OH
MW: 62.07 g/mol
Boiling Pt: 196…198 °C (760 mmHg)
Melting Pt: -13
Density: 1.113 g/cm³
Flash Pt: 116 °C
Storage Temperature: Ambient
MDL Number: MFCD00002885
CAS Number: 107-21-1
EINECS: 203-473-3
REACH: 01-2119456816-28
Merck Index: 13,03832
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c4d1c97392851ba184f445f6ee912f84631f98d9&vid=2002300f174929a030d66d9f1394eb6bee89b7dc&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Clear viscous liquid
Infrared spectrum Conforms
Water ≤0.1 % (K.F.)
Sulfated ash ≤0.005 %
GC ≥99.5 %
Color scale ≤10 APHA
Acidity ≤0.0002 meq/g
Chloride (Cl) ≤0.2 ppm
Iron (Fe) ≤0.5 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR