Cesium carbonate 99.99% Trace metals basis

Supplier: Thermo Scientific Chemicals
Danger

AC27802-0100 AC27802-0500
TS27802-0100EA 48 USD
TS27802-0100 TS27802-0500
Cesium carbonate 99.99% Trace metals basis
Cesium carbonate
Formula: Cs₂CO₃
MW: 325.82 g/mol
Melting Pt: 610
Storage Temperature: Ambient
MDL Number: MFCD00010957
CAS Number: 534-17-8
EINECS: 208-591-9
Merck Index: 13,02020

Order Now

Specification Test Results

Appearance White Powder
Total trace metal impurities ≤0.01 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR