Fuchsin (acid), pure, high purity biological stain

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Benzenesulfonic acid, 2-amino-5-[(4-amino-3-sulfophenyl)(4-imino-3-sulfo-2,5-cyclohexadien-1-ylidene)methyl]-3-methyl-, sodium salt (1:2), Rubin S, CN DcR-8, Acid Violet 19, 3-(1-(4-Amino-3-methyl-5-sulfonatophenyl)-1-(4-amino-3-sulfonatophenyl)methylene)cyclohexa-1,4-dienesulfonic acid, Acid Fuchsin, Acid Fuchsin sodium salt, Fuchsin S, CI 42685, Acid Magenta, 3-(1-(4-Amino-3-methyl-5-sulphonatophenyl)-1-(4-amino-3-sulphonatophenyl)methylene)cyclohexa-1,4-dienesulphonic acid, Benzenesulphonic acid, 2-amino-5-[(4-amino-3-sulphophenyl)(4-imino-3-sulpho-2,5-cyclohexadien-1-ylidene)methyl]-3-methyl-, sodium salt (1:2)

22790-1000 22790-0250 22790-5000
TS22790-1000EA 229 USD
TS22790-1000 TS22790-0250 TS22790-5000
Fuchsin (acid), pure, high purity biological stain
Fuchsin (acid)
Formula: C₂₀H₁₇N₃Na₂O₉S₃
Storage Temperature: Ambient
MDL Number: MFCD00013286
CAS Number: 3244-88-0
EINECS: 221-816-5
Merck Index: 12,00109

Order Now

Specification Test Results

Appearance Dark green Powder or crystals
Infrared spectrum Conforms
UV Lambda max 544 to 550 nm (in pH 3 acidified water)
Absorption Ratio P-15/P+15 1 to 1.4
Specific absorbance A (1%/1cm) ≥700 (at lambda max.)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR