Calcium chloride 96%, granules for analysis

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Chloro calcium, Calcium dichloride, CaCl2

AC21917-0025 AC21917-1000 AC21917-5000
TS21917-0025EA 209 USD
TS21917-0025 TS21917-1000 TS21917-5000
Calcium chloride 96%, granules for analysis
Calcium chloride
Formula: CaCl₂
Boiling Pt: >1600 °C (760 mmHg)
Melting Pt: 782 °C
Storage Temperature: Ambient
MDL Number: MFCD00010903
CAS Number: 10043-52-4

Order Now

Specification Test Results

Appearance White Granules
Titration Complexometric ≥95 %
Fluoride (F) ≤40 ppm
Sulfate (SO4) ≤0.2 %
Arsenic (As) ≤1 ppm
Barium (Ba) ≤0.4 %
Iron (Fe) ≤50 ppm
Lead (Pb) ≤5 ppm
Mercury (Hg) ≤1 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR