n-Pentane, anhydrous 99+%, extra pure

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Pentane

217240010 217240025 217240251
TS217240010EA 114 USD
TS217240010 TS217240025 TS217240251
n-Pentane, anhydrous 99+%, extra pure
n-Pentane
Formula: H₃C(CH₂)₃CH₃
MW: 72.15 g/mol
Boiling Pt: 36 °C (760 mmHg)
Melting Pt: -130
Density: 0.626 g/cm³
Flash Pt: –49 °C
Storage Temperature: Ambient
MDL Number: MFCD00009498
CAS Number: 109-66-0
EINECS: 203-692-4
UN: 1265
ADR: 3 II
REACH: 01-2119459286-30
Merck Index: 13,07193

Order Now

Specification Test Results

Appearance Clear liquid
Infrared spectrum Conforms
Refractive index 1.3570 to 1.3590 (20°C, 589 nm)
Water ≤0.005 %
GC ≥99.0 %
Color scale ≤10 APHA
Residue after evaporation ≤0.001 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR