Iodine monochloride 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Chloroiodide, Iodochloride, Iodine chloride

AC19696-1000 AC19696-5000
TS19696-1000EA 137 USD
TS19696-1000 TS19696-5000
Iodine monochloride 98%
Iodine monochloride
Formula: ClI
Boiling Pt: 97 °C (760 mmHg)
Melting Pt: 26 °C
Density: 3.2 g/cm³
Storage Temperature: Refrigerator
MDL Number: MFCD00011354
CAS Number: 7790-99-0
EINECS: 232-236-7
UN: 1792
ADR: 8,II
Merck Index: 13,05039

Order Now

Specification Test Results

Appearance Dark red-brown to brown to black Liquid or solid
Titration Iodometric ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR