2-Ethoxyethanol 99%, extra pure

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Ethyl glycol, Ethylene glycol monoethyl ether, Cellosolve

15602-0010 156020025 156020250
TS15602-0010EA 76 USD
TS15602-0010 TS156020025 TS156020250
2-Ethoxyethanol 99%, extra pure
2-Ethoxyethanol
Formula: C₂H₅OCH₂CH₂OH
Boiling Pt: 135 °C (760 mmHg)
Melting Pt: –70 °C
Density: 0.93 g/cm³
Flash Pt: 40 °C
Storage Temperature: Ambient
MDL Number: MFCD00002869
CAS Number: 110-80-5
EINECS: 203-804-1
UN: 1171
ADR: 3,III
REACH: 01-2119560582-38
Merck Index: 13,03786
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=461a4daf8709ebd6b4516d73b4b32eee33cd1bca&vid=625adefb8d1e2126363931c57a51d3e1df419926&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Clear liquid
Infrared spectrum Conforms
Refractive index 1.4060 to 1.4100 (20°C, 589 nm)
Water ≤0.15 % (K.F.)
GC ≥98.5 %
Color scale ≤10 APHA
Specific gravity (20°C) 0.928 to 0.932
Alkalinity ≤0.0002 meq/g
Substance darkened by H2SO4 passes test
Calcium (Ca) ≤0.5 ppm
Chromium (Cr) ≤0.02 ppm
Cobalt (Co) ≤0.02 ppm
Copper (Cu) ≤0.05 ppm
Iron (Fe) ≤0.2 ppm
Magnesium (Mg) ≤0.1 ppm
Manganese (Mn) ≤0.02 ppm
Nickel (Ni) ≤0.02 ppm
Zinc (Zn) ≤0.1 ppm
Heavy metals (as Pb) ≤1 ppm
Ethanol 99.8 to 100 % v/v (calculated)
Substance reducing dichromate passes test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR