(Methoxymethyl)triphenylphosphonium chloride 98%

Supplier: Thermo Scientific Chemicals
Warning

14601-0250 14601-1000 14601-5000
TS14601-0250EA 67 USD
TS14601-0250 TS14601-1000 TS14601-5000
(Methoxymethyl)triphenylphosphonium chloride 98%
(Methoxymethyl)triphenylphosphonium chloride
Formula: C₂₀H₂₀ClOP
Boiling Pt: 413 °C
Melting Pt: 195…197 °C
Flash Pt: >250 °C
Storage Temperature: Ambient
MDL Number: MFCD00011800
CAS Number: 4009-98-7
EINECS: 223-664-5
UN: 3077
ADR: 9,III

Order Now

Specification Test Results

Appearance White to almost white Powder or crystals
Infrared spectrum Conforms
Water ≤0.5 %
HPLC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR